siRNA Details
| Gene Symbol: Mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004233.1 | NP_001004233.1 | 2 | 227 | ||
| XM_006236301.2 | XP_006236363.1 | 2 | 427 | |||
| XM_006236303.2 | XP_006236365.1 | 2 | 383 | |||
| XM_006236304.2 | XP_006236366.1 | 2 | 243 | |||
| GenBank mRNA | BC079407.1 | - | 2 | 227 | 7112867 | |
| FQ226725.1 | - | 2 | 378 | |||
| FQ233779.1 | - | 2 | 376 | |||

Mkrn1
Genomic Map