siRNA Details
| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181371.2 | NP_852036.1 | 3, 4 | 315 | ||
| GenBank mRNA | FJ179400.1 | - | 3, 4 | 281 | ||
| FQ211030.1 | - | 3, 4 | 333 | |||
| FQ214179.1 | - | 3, 4 | 398 | |||
| FQ220456.1 | - | 3, 4 | 371 | |||
| S83436.1 | - | 3, 4 | 315 | |||

Gstk1
Genomic Map