siRNA Details
| Gene Symbol: Gimap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011968.3 | NP_001011968.1 | 3 | 972 | ||
| GenBank mRNA | AJ633683.1 | - | 3 | 975 | ||
| BC089859.1 | - | 881 | 7384582 | |||
| DQ125342.1 | - | 3 | 882 | |||
| DQ125343.1 | - | 882 | ||||

Gimap6
Genomic Map