siRNA Details
| Gene Symbol: Svs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199095.1 | NP_954526.1 | 1 | 1527 | ||
| XM_006236459.1 | XP_006236521.1 | 2 | 1269 | |||
| GenBank mRNA | D00921.1 | - | 1 | 760 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Svs1| Gene Symbol: Svs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199095.1 | NP_954526.1 | 1 | 1527 | ||
| XM_006236459.1 | XP_006236521.1 | 2 | 1269 | |||
| GenBank mRNA | D00921.1 | - | 1 | 760 | ||