siRNA Details
| Gene Symbol: Svs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199095.1 | NP_954526.1 | 4 | 2711 | ||
| XM_006236459.1 | XP_006236521.1 | 5 | 2453 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Svs1| Gene Symbol: Svs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199095.1 | NP_954526.1 | 4 | 2711 | ||
| XM_006236459.1 | XP_006236521.1 | 5 | 2453 | |||