siRNA Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006965.3 | NP_008896.2 | 4 | 1227 | ||
| XM_005258341.3 | XP_005258398.1 | 4 | 1214 | |||
| GenBank mRNA | AF038964.1 | - | 4 | 1171 | ||
| AK291246.1 | - | 4 | 1206 | |||
| BC003566.1 | - | 4 | 1177 | 3537160 | ||
| BC016801.1 | - | 4 | 1198 | 4072599 | ||
| BT006658.1 | - | 3 | 1036 | |||
| CR450328.1 | - | 3 | 1036 | |||
| U68536.1 | - | 4 | 1200 | |||
| X52348.1 | - | 1 | 82 | |||

ZNF24
Genomic Map