siRNA Details
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007646.1 | NP_001007647.1 | 4 | 686 | ||
GenBank mRNA | BC081961.1 | - | 4 | 686 | 7124723 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007646.1 | NP_001007647.1 | 4 | 686 | ||
GenBank mRNA | BC081961.1 | - | 4 | 686 | 7124723 |