siRNA Details
Gene Symbol: Mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106596.1 | NP_001100066.1 | 4 | 486 | ||
XM_006236635.1 | XP_006236697.1 | 4 | 462 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106596.1 | NP_001100066.1 | 4 | 486 | ||
XM_006236635.1 | XP_006236697.1 | 4 | 462 |