siRNA Details
| Gene Symbol: Capg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013086.1 | NP_001013104.1 | 9 | 1259 | ||
| XM_006236661.2 | XP_006236723.1 | 10 | 1099 | |||
| XM_006236662.2 | XP_006236724.1 | 10 | 1192 | |||
| XM_006236663.2 | XP_006236725.1 | 10 | 1326 | |||
| XM_006236666.2 | XP_006236728.1 | 9 | 1046 | |||
| GenBank mRNA | BC079104.1 | - | 9 | 1259 | 7125038 | |

Capg
Genomic Map