siRNA Details
| Gene Symbol: Htra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106599.2 | NP_001100069.1 | 3, 4 | 923 | ||
| GenBank mRNA | BC087588.1 | - | 3, 4 | 903 | ||
| BC158760.1 | - | 3, 4 | 923 | 7103492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Htra2| Gene Symbol: Htra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106599.2 | NP_001100069.1 | 3, 4 | 923 | ||
| GenBank mRNA | BC087588.1 | - | 3, 4 | 903 | ||
| BC158760.1 | - | 3, 4 | 923 | 7103492 | ||