siRNA Details
Gene Symbol: Ccdc142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_232111.7 | XP_232111.4 | 8, 9 | 3695 |
Gene Symbol: LOC103692170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763085.1 | XP_008761307.1 | 3695 |
