siRNA Details
| Gene Symbol: LOC103692170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763085.1 | XP_008761307.1 | 3357 | |||
| Gene Symbol: Ccdc142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_232111.7 | XP_232111.4 | 7 | 3357 | ||

LOC103692170
Genomic Map