siRNA Details
| Gene Symbol: Mob1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033891.1 | NP_001029063.1 | 5 | 651 | ||
| GenBank mRNA | BC101879.1 | - | 5 | 651 | 7929996 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mob1a| Gene Symbol: Mob1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033891.1 | NP_001029063.1 | 5 | 651 | ||
| GenBank mRNA | BC101879.1 | - | 5 | 651 | 7929996 | |