siRNA Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017537.1 | NP_001017537.1 | 3, 4 | 331 | ||
| XM_008763059.1 | XP_008761281.1 | 3, 4 | 409 | |||
| XM_008763060.1 | XP_008761282.1 | 3, 4 | 537 | |||
| XM_008763061.1 | XP_008761283.1 | 3, 4 | 284 | |||
| GenBank mRNA | BC091302.1 | - | 3, 4 | 331 | 7309682 | |
| FQ214220.1 | - | 3, 4 | 343 | |||
| FQ218231.1 | - | 3, 4 | 342 | |||

Tex261
Genomic Map