siRNA Details
| Gene Symbol: Nagk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037768.1 | NP_001032857.1 | 8 | 737 | ||
| GenBank mRNA | BC107647.1 | - | 8 | 737 | 7930414 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nagk| Gene Symbol: Nagk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037768.1 | NP_001032857.1 | 8 | 737 | ||
| GenBank mRNA | BC107647.1 | - | 8 | 737 | 7930414 | |