siRNA Details
| Gene Symbol: Nagk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037768.1 | NP_001032857.1 | 5 | 427 | ||
| XM_006236740.2 | XP_006236802.1 | 5 | 1483 | |||
| GenBank mRNA | BC079456.1 | - | 5 | 409 | ||
| BC107647.1 | - | 5 | 427 | 7930414 | ||

Nagk
Genomic Map