siRNA Details
| Gene Symbol: Cct7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106603.1 | NP_001100073.1 | 5, 6 | 1089 | ||
| GenBank mRNA | FQ214319.1 | - | 5, 6 | 459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct7| Gene Symbol: Cct7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106603.1 | NP_001100073.1 | 5, 6 | 1089 | ||
| GenBank mRNA | FQ214319.1 | - | 5, 6 | 459 | ||