siRNA Details
| Gene Symbol: Gkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198972.1 | NP_945323.1 | 3, 4 | 235 | ||
| GenBank mRNA | AY456958.1 | - | 3, 4 | 235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gkn1| Gene Symbol: Gkn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198972.1 | NP_945323.1 | 3, 4 | 235 | ||
| GenBank mRNA | AY456958.1 | - | 3, 4 | 235 | ||