siRNA Details
| Gene Symbol: Abtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005902.1 | NP_001005902.1 | 2 | 168 | ||
| GenBank mRNA | BC083579.1 | - | 2 | 168 | 7128126 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abtb1| Gene Symbol: Abtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005902.1 | NP_001005902.1 | 2 | 168 | ||
| GenBank mRNA | BC083579.1 | - | 2 | 168 | 7128126 | |