siRNA Details
| Gene Symbol: Chchd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106608.1 | NP_001100078.1 | 4 | 572 | ||
| XM_006236846.2 | XP_006236908.1 | 4 | 579 | |||
| GenBank mRNA | FQ219980.1 | - | 4 | 553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chchd6| Gene Symbol: Chchd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106608.1 | NP_001100078.1 | 4 | 572 | ||
| XM_006236846.2 | XP_006236908.1 | 4 | 579 | |||
| GenBank mRNA | FQ219980.1 | - | 4 | 553 | ||