siRNA Details
Gene Symbol: Vom1r100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173296.1 | NP_775418.1 | 3 | 765 | ||
GenBank mRNA | AY510294.1 | - | 1 | 630 | ||
U36895.1 | - | 3 | 765 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vom1r100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173296.1 | NP_775418.1 | 3 | 765 | ||
GenBank mRNA | AY510294.1 | - | 1 | 630 | ||
U36895.1 | - | 3 | 765 |