siRNA Details
| Gene Symbol: Vom1r100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173296.1 | NP_775418.1 | 3 | 1015 | ||
| GenBank mRNA | U36895.1 | - | 3 | 1015 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r100| Gene Symbol: Vom1r100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173296.1 | NP_775418.1 | 3 | 1015 | ||
| GenBank mRNA | U36895.1 | - | 3 | 1015 | ||