siRNA Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236885.2 | XP_006236947.1 | 8 | 1520 | ||
| XM_006236886.2 | XP_006236948.1 | 7 | 1344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc37| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236885.2 | XP_006236947.1 | 8 | 1520 | ||
| XM_006236886.2 | XP_006236948.1 | 7 | 1344 | |||