siRNA Details
| Gene Symbol: Ccdc174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009659.2 | NP_001009659.2 | 5 | 475 | ||
| XM_006236907.2 | XP_006236969.1 | 5 | 512 | |||
| GenBank mRNA | BC087050.1 | - | 5 | 475 | 7114831 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc174| Gene Symbol: Ccdc174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009659.2 | NP_001009659.2 | 5 | 475 | ||
| XM_006236907.2 | XP_006236969.1 | 5 | 512 | |||
| GenBank mRNA | BC087050.1 | - | 5 | 475 | 7114831 | |