siRNA Details
| Gene Symbol: ZNF33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278170.1 | NP_001265099.1 | 4 | 2433 | ||
| NM_001278171.1 | NP_001265100.1 | 6 | 2612 | |||
| NM_001278173.1 | NP_001265102.1 | 6 | 2568 | |||
| NM_001278174.1 | NP_001265103.1 | 6 | 2564 | |||
| NM_001278175.1 | NP_001265104.1 | 6 | 2553 | |||
| NM_001278176.1 | NP_001265105.1 | 6 | 2569 | |||
| NM_001278177.1 | NP_001265106.1 | 5 | 2485 | |||
| NM_001278178.1 | NP_001265107.1 | 5 | 2494 | |||
| NM_001278179.1 | NP_001265108.1 | 6 | 2609 | |||
| NM_001324175.1 | NP_001311104.1 | 6 | 2561 | |||
| NM_001324176.1 | NP_001311105.1 | 5 | 2491 | |||
| NM_001324177.1 | NP_001311106.1 | 6 | 2569 | |||
| NM_001324178.1 | NP_001311107.1 | 4 | 2436 | |||
| NM_006954.1 | NP_008885.1 | 5 | 2489 | |||
| NM_006974.2 | NP_008905.1 | 5 | 2486 | |||
| XM_011519650.2 | XP_011517952.1 | 6 | 2558 | |||
| XM_011519651.2 | XP_011517953.1 | 6 | 2555 | |||
| XM_011519653.2 | XP_011517955.1 | 4 | 2371 | |||
| XM_011519654.2 | XP_011517956.1 | 4 | 2368 | |||
| XM_017016617.1 | XP_016872106.1 | 5 | 2452 | |||
| GenBank mRNA | AK292844.1 | - | 5 | 2470 | ||
| AK303445.1 | - | 4 | 2428 | |||
| AY184389.1 | - | 3 | 2308 | |||
| BC126119.1 | - | 3 | 2361 | 8991835 | ||
| D31763.1 | - | 5 | 2423 | |||
| X68687.1 | - | 1 | 426 | |||

ZNF33A
Genomic Map