siRNA Details
| Gene Symbol: Crbn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015003.2 | NP_001015003.1 | 5 | 565 | ||
| GenBank mRNA | AY973953.1 | - | 5 | 552 | ||
| BC099779.1 | - | 5 | 550 | |||
| BC127455.1 | - | 5 | 557 | 7112865 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crbn| Gene Symbol: Crbn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015003.2 | NP_001015003.1 | 5 | 565 | ||
| GenBank mRNA | AY973953.1 | - | 5 | 552 | ||
| BC099779.1 | - | 5 | 550 | |||
| BC127455.1 | - | 5 | 557 | 7112865 | ||