siRNA Details
| Gene Symbol: Sec13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006978.1 | NP_001006979.1 | 6 | 552 | ||
| GenBank mRNA | BC084705.1 | - | 6 | 552 | 7301091 | |
| FQ212944.1 | - | 6 | 572 | |||
| FQ216715.1 | - | 6 | 570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec13| Gene Symbol: Sec13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006978.1 | NP_001006979.1 | 6 | 552 | ||
| GenBank mRNA | BC084705.1 | - | 6 | 552 | 7301091 | |
| FQ212944.1 | - | 6 | 572 | |||
| FQ216715.1 | - | 6 | 570 | |||