siRNA Details
| Gene Symbol: Vgll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015004.1 | NP_001015004.1 | 2 | 596 | ||
| XM_008763228.1 | XP_008761450.1 | 2 | 1152 | |||
| XM_008763229.1 | XP_008761451.1 | 3 | 503 | |||
| XM_008763230.1 | XP_008761452.1 | 3 | 369 | |||
| GenBank mRNA | BC091399.1 | - | 2 | 596 | 7323935 | |
| FQ220379.1 | - | 3 | 282 | |||

Vgll4
Genomic Map