siRNA Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008314.1 | NP_001008315.1 | 8 | 1153 | ||
| GenBank mRNA | BC083870.1 | - | 8 | 1153 | 7128406 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkrn2| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008314.1 | NP_001008315.1 | 8 | 1153 | ||
| GenBank mRNA | BC083870.1 | - | 8 | 1153 | 7128406 | |