siRNA Details
| Gene Symbol: Clec4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005880.1 | NP_001005880.1 | 3 | 309 | ||
| GenBank mRNA | AY397670.1 | - | 3 | 309 | ||
| AY581052.1 | - | 3 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4a2| Gene Symbol: Clec4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005880.1 | NP_001005880.1 | 3 | 309 | ||
| GenBank mRNA | AY397670.1 | - | 3 | 309 | ||
| AY581052.1 | - | 3 | 309 | |||