siRNA Details
| Gene Symbol: Spsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009660.1 | NP_001009660.1 | 2 | 380 | ||
| XM_006237336.2 | XP_006237398.1 | 2 | 869 | |||
| XM_006237337.2 | XP_006237399.1 | 2 | 416 | |||
| GenBank mRNA | BC088189.1 | - | 2 | 380 | 7371446 | |
| Gene Symbol: LOC100911548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750655.3 | XP_003750703.1 | 2 | 492 | ||
| XM_006244625.2 | XP_006244687.1 | 2 | 865 | |||
| XM_006244626.2 | XP_006244688.1 | 2 | 414 | |||

Spsb2
Genomic Map