siRNA Details
| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106622.1 | NP_001100092.1 | 6 | 1618 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tapbpl| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106622.1 | NP_001100092.1 | 6 | 1618 | ||