siRNA Details
| Gene Symbol: Klra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198746.1 | NP_942041.1 | 7 | 878 | ||
| GenBank mRNA | U56863.1 | - | 7 | 878 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra5| Gene Symbol: Klra5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198746.1 | NP_942041.1 | 7 | 878 | ||
| GenBank mRNA | U56863.1 | - | 7 | 878 | ||