siRNA Details
| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015005.2 | NP_001015005.2 | 5 | 543 | ||
| XM_006237518.1 | XP_006237580.1 | 6 | 600 | |||
| GenBank mRNA | BC091696.1 | - | 5 | 536 | 7301732 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx47| Gene Symbol: Ddx47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015005.2 | NP_001015005.2 | 5 | 543 | ||
| XM_006237518.1 | XP_006237580.1 | 6 | 600 | |||
| GenBank mRNA | BC091696.1 | - | 5 | 536 | 7301732 | |