siRNA Details
| Gene Symbol: Plbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013927.1 | NP_001013949.1 | 2 | 318 | ||
| GenBank mRNA | BC085851.1 | - | 2 | 318 | 7189866 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plbd1| Gene Symbol: Plbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013927.1 | NP_001013949.1 | 2 | 318 | ||
| GenBank mRNA | BC085851.1 | - | 2 | 318 | 7189866 | |