siRNA Details
| Gene Symbol: Casc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106627.3 | NP_001100097.2 | 14, 15 | 1847 | ||
| XM_008763378.1 | XP_008761600.1 | 10, 11 | 2284 | |||
| GenBank mRNA | BC160907.1 | - | 14, 15 | 1847 | 7455489 | |
| DV722478.1 | - | 2, 3 | 334 | |||

Casc1
Genomic Map