siRNA Details
| Gene Symbol: Mrps35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106628.1 | NP_001100098.1 | 5, 6 | 551 | ||
| XM_006237676.1 | XP_006237738.1 | 5, 6 | 556 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps35| Gene Symbol: Mrps35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106628.1 | NP_001100098.1 | 5, 6 | 551 | ||
| XM_006237676.1 | XP_006237738.1 | 5, 6 | 556 | |||