siRNA Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024984.1 | NP_001020155.1 | 2, 3 | 202 | ||
| GenBank mRNA | BC083597.1 | - | 2, 3 | 202 | 7132841 | |
| FQ213743.1 | - | 2, 3 | 184 | |||
| FQ227558.1 | - | 2, 3 | 183 | |||
| FQ229098.1 | - | 2, 3 | 203 | |||
| FQ230170.1 | - | 2, 3 | 204 | |||

Ergic2
Genomic Map