siRNA Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024984.1 | NP_001020155.1 | 4 | 379 | ||
| GenBank mRNA | BC083597.1 | - | 4 | 379 | 7132841 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ergic2| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024984.1 | NP_001020155.1 | 4 | 379 | ||
| GenBank mRNA | BC083597.1 | - | 4 | 379 | 7132841 | |