siRNA Details
| Gene Symbol: Terf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012464.1 | NP_001012482.1 | 8, 9 | 1008 | ||
| XM_006237729.2 | XP_006237791.1 | 7, 8 | 981 | |||
| GenBank mRNA | BC089888.1 | - | 8, 9 | 1008 | 7374057 | |

Terf1
Genomic Map