siRNA Details
| Gene Symbol: Xkr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011971.1 | NP_001011971.1 | 4 | 1633 | ||
| GenBank mRNA | AY534258.1 | - | 4 | 1633 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr4| Gene Symbol: Xkr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011971.1 | NP_001011971.1 | 4 | 1633 | ||
| GenBank mRNA | AY534258.1 | - | 4 | 1633 | ||