siRNA Details
| Gene Symbol: Sdr16c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106634.1 | NP_001100104.1 | 4 | 831 | ||
| XM_006237836.2 | XP_006237898.1 | 5 | 718 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdr16c5| Gene Symbol: Sdr16c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106634.1 | NP_001100104.1 | 4 | 831 | ||
| XM_006237836.2 | XP_006237898.1 | 5 | 718 | |||