siRNA Details
| Gene Symbol: Hdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025409.1 | NP_001020580.1 | 5, 6 | 513 | ||
| GenBank mRNA | BC059156.1 | - | 5, 6 | 530 | ||
| BC097943.1 | - | 5, 6 | 513 | 7458532 | ||
| BC107476.1 | - | 5, 6 | 510 | 7443793 | ||

Hdac1
Genomic Map