siRNA Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024757.1 | NP_001019928.1 | 19 | 2335 | ||
| XM_006237930.2 | XP_006237992.1 | 19 | 2250 | |||
| XM_006237931.2 | XP_006237993.1 | 18 | 2677 | |||
| XM_008763590.1 | XP_008761812.1 | 20 | 2548 | |||
| XM_008763591.1 | XP_008761813.1 | 12 | 1743 | |||
| GenBank mRNA | BC097386.1 | - | 19 | 2335 | 7446512 | |
| FQ216775.1 | - | 4 | 426 | |||

Wwp1
Genomic Map