siRNA Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024757.1 | NP_001019928.1 | 4 | 432 | ||
| XM_006237930.2 | XP_006237992.1 | 4 | 347 | |||
| XM_006237931.2 | XP_006237993.1 | 3 | 774 | |||
| XM_008763590.1 | XP_008761812.1 | 5 | 645 | |||
| GenBank mRNA | BC097386.1 | - | 4 | 432 | 7446512 | |

Wwp1
Genomic Map