siRNA Details
| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025274.2 | NP_001020445.2 | 11 | 2278 | ||
| GenBank mRNA | BC091301.1 | - | 2 | 1357 | 7309546 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnisr| Gene Symbol: Pnisr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025274.2 | NP_001020445.2 | 11 | 2278 | ||
| GenBank mRNA | BC091301.1 | - | 2 | 1357 | 7309546 | |