siRNA Details
| Gene Symbol: Smim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106644.1 | NP_001100114.1 | 3 | 568 | ||
| NM_001201374.1 | NP_001188303.1 | 2 | 190 | |||
| NM_001201375.1 | NP_001188304.1 | 2 | 434 | |||
| NM_001201376.1 | NP_001188305.1 | 2 | 183 | |||
| XM_006237987.2 | XP_006238049.1 | 3 | 473 | |||
| XM_006237988.2 | XP_006238050.1 | 2 | 356 | |||
| GenBank mRNA | DY544415.1 | - | 2 | 190 | ||
| FM105726.1 | - | 2 | 434 | |||
| FM134958.1 | - | 2 | 183 | |||
| FQ226097.1 | - | 2 | 395 | |||

Smim8
Genomic Map