siRNA Details
| Gene Symbol: Chmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025410.1 | NP_001020581.1 | 5 | 372 | ||
| GenBank mRNA | BC086333.1 | - | 4 | 234 | ||
| BC097963.1 | - | 5 | 372 | 7387550 | ||
| FQ214132.1 | - | 5 | 400 | |||
| FQ228147.1 | - | 5 | 406 | |||
| FQ234681.1 | - | 5 | 406 | |||

Chmp5
Genomic Map