siRNA Details
| Gene Symbol: Nudt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207596.1 | NP_997479.1 | 3 | 196 | ||
| GenBank mRNA | BC058156.1 | - | 3 | 196 | 6888562 | |
| FQ227408.1 | - | 3 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nudt2| Gene Symbol: Nudt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207596.1 | NP_997479.1 | 3 | 196 | ||
| GenBank mRNA | BC058156.1 | - | 3 | 196 | 6888562 | |
| FQ227408.1 | - | 3 | 197 | |||