siRNA Details
| Gene Symbol: Ccl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008513.1 | NP_001008513.1 | 2 | 170 | ||
| GenBank mRNA | BC086571.1 | - | 2 | 170 | 7304213 | |
| FQ218040.1 | - | 2 | 194 | |||
| FQ220091.1 | - | 2 | 194 | |||
| FQ223815.1 | - | 2 | 196 | |||
| FQ225478.1 | - | 2 | 194 | |||
| FQ226079.1 | - | 2 | 192 | |||
| FQ233622.1 | - | 2 | 194 | |||

Ccl21
Genomic Map